Learn More
Streptozotocin, 97+%, Thermo Scientific™
Streptozotocin (streptozocin, CAS # 18883-66-4) is an N-nitrosourea antibiotic produced by Streptomyces achromogenes.
164.00€ - 280.99€
Chemical Identifiers
| CAS | 18883-66-4 |
|---|---|
| Molecular Formula | C8H15N3O7 |
| Molecular Weight (g/mol) | 265.222 |
| MDL Number | MFCD00006607 |
| InChI Key | ZSJLQEPLLKMAKR-UHEPRFFZSA-N |
| Synonym | NSC-85998; U-9889 |
| PubChem CID | 45357367 |
| IUPAC Name | 1-methyl-1-nitroso-3-[(2R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea |
| SMILES | CN(C(=O)NC1C(C(C(OC1O)CO)O)O)N=O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15434179
|
Thermo Scientific Alfa Aesar
J61601.ME |
500mg |
167.00€ 164.00€ / 500mg Save 3.00€ 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15424179
|
Thermo Scientific Alfa Aesar
J61601.03 |
1g |
287.00€ 280.99€ / 1g Save 6.01€ 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Streptozotocin acts as a bactericidal by alkylating DNA and inhibiting DNA synthesis
• Streptozotocin enters cells through glucose transporter 2
Application
• Streptozotocin induces diabetes in experimental animals and can cause acute liver injury
• It is also useful for research on neuroendocrine tumors and carcinoma cells
Chemical Identifiers
| 18883-66-4 | |
| 265.222 | |
| ZSJLQEPLLKMAKR-UHEPRFFZSA-N | |
| 45357367 | |
| CN(C(=O)NC1C(C(C(OC1O)CO)O)O)N=O |
| C8H15N3O7 | |
| MFCD00006607 | |
| NSC-85998; U-9889 | |
| 1-methyl-1-nitroso-3-[(2R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea |
Specifications
| 18883-66-4 | |
| Hygroscopic | |
| MFCD00006607 | |
| 14,8832 | |
| Soluble in water or 100% ethanol | |
| CN(C(=O)NC1C(C(C(OC1O)CO)O)O)N=O | |
| 265.222 | |
| 265.22 | |
| Streptozotocin |
| White | |
| C8H15N3O7 | |
| 2060675 | |
| NSC-85998; U-9889 | |
| ZSJLQEPLLKMAKR-UHEPRFFZSA-N | |
| 1-methyl-1-nitroso-3-[(2R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea | |
| 45357367 | |
| ≥97% |
Safety and Handling
GHS H Statement
H351
Suspected of causing cancer.
P201-P202-P281-P308+P313-P501c
H350
missing translation for 'einecsNumber' : 242-646-8
missing translation for 'rtecsNumber' : LZ5775000
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.