missing translation for 'onlineSavingsMsg'
Learn More

Strontium 2-ethylhexanoate, 40% in 2-ethylhexanoic acid (8-12% Sr), Thermo Scientific Chemicals

Catalog Number p-7051198
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2457-02-5
MFCD00070438
This item is not returnable. View return policy
2457-02-5
MFCD00070438

Strontium 2-ethylhexanoate is used in metal refining, plumbing, bleaching, engraving, plating, photography, disinfection, fertilizer manufacture, metal cleaning, and rust removal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Odor Mild
Molecular Formula C16H30O4Sr
Quantity 5 g
Synonym strontium 2-ethylhexanoate, strontium bis 2-ethylhexanoate, hexanoic acid, 2-ethyl-, strontium salt 2:1, acmc-20ajmv, bis 2-ethylhexanoic acid strontium salt, hexanoic acid, 2-ethyl-, strontium salt, strontium 2+ bis 2-ethylhexanoate, hexanoic acid,2-ethyl-, strontium salt 2:1
Solubility Information Insoluble in water.
InChI Key DLWBHRIWCMOQKI-UHFFFAOYNA-L
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sr+2]
IUPAC Name strontium;2-ethylhexanoate
Molecular Weight (g/mol) 374.03
PubChem CID 102822
Formula Weight 374.04
Concentration or Composition (by Analyte or Components) 40% in 2-ethylhexanoic acid (8 to 12% Sr)
Physical Form Liquid
Chemical Name or Material Strontium 2-ethylhexanoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'