Learn More
Cole-Parmer™ Dual Tube Holder Disc Adapter for Cole-Parmer™ TR-200 Series Tube Rotators
Product Code. 11486548
Change view
Click to view available options
For Use With (Equipment):
Stuart™ SB2/SB3 Tube Rotator Holders
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
11486548 Stuart™ SB2/SB3 Tube Rotator Holders Each
1 options
Product Code. 11486548
Brand: Cole-Parmer™ 0007723
Only null left
Add to basket
Add to basket
The Dual Tube Holder Disc Adapter for Cole-Parmer™ TR-200 Series Tube Rotators saves time by rotating twice the amount. Affixes easily without tools. Fits two tube holders to the SB2 or SB3 Tube Rotator simultaneously.
  • Choose the tube holder(s) that accommodates your tube size(s).
  • Fits two tube holders to Cole-Parmer™ TR-200 series tube rotator simultaneously.
  • 1 year warranty.
Specifications
Product Type Adapter
For Use With (Equipment) Stuart™ SB2/SB3 Tube Rotator Holders