Learn More
Cole-Parmer™ Tube Holder Disc for Cole-Parmer™ TR-200 Series Tube Rotators
Product Code. p-8012889
Click to view available options
For Use With (Equipment):
For 12 Tubes
For 20 Glaser
For 30 Tubes, 26 mm dia.
For 40 Microcentrifuge Tubes
For 63 Tubes, 12 mm dia.
For 63 Tubes, 16 mm dia.
Unit Size:
Each
Récupel-bijdrage uitgesloten
Product Code. 12027144
Brand: Cole-Parmer™ 0007713
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Quickly and easily replace tube holder without tools using these Tube Holder Discs for Cole-Parmer™ TR-200 Series Tube Rotators.
  • Choose the tube holder(s) that accommodates your tube size(s).
Specifications
Holds Centrifuge tubes
Tube Capacity 12 x 50 mL
Product Type Tube Holder
For Use With (Equipment) For 12 Tubes
No. of Positions 12 Tubes