Learn More
Cole-Parmer™ Stuart™ Heater Block Inserts
Product Code. p-7182442
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
Size:
Large Base
Medium Base
Small and Medium Bases
Unit Size:
Each
Product Code. 15143027
Brand: Cole-Parmer™ 0480597
Only null left
Add to basket
Add to basket
Accessory heating block for 100 mL round bottom flasks for use with the Stuart™ SHB1000BASE and Stuart™ Undergrad hotplate
  • Safest way to heat round-bottom flasks
  • Flexible modular system for a wide range of sizes up to 500 mL
  • Low-well design is perfect for multi-neck flasks
Specifications
Capacity (Metric) 50 mL
Holds 50 ml tubes
Material Anodized Aluminum
No. of Places 3
Type Block
Quantity 3/Pk.
Size Small and Medium Bases