Learn More
Cole-Parmer™ Stuart™ Platform
Product Code. 11476608
Change view
Click to view available options
Dimensions (L x W):
340 x 300 mm
510 x 510 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
11476608 510 x 510 mm Each
11456608 340 x 300 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Récupel-bijdrage uitgesloten
Product Code. 11476608 Supplier Cole-Parmer™ Supplier No. 5170095
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Holds Erhlenmeyer Flasks or Bottles using rubber securing bars. Stuart™ Platforms with flexible cradle system allows different combinations of vessels to be used, offering maximum versatility. For use with Stuart™ Orbital Shakers (SSL1, SSM1)
  • For use with Stuart&;trade; Orbital Shakers (SSL1, SSM1)
  • Holds up Erhlenmeyer Flasks or Bottles using rubber securing bars.
  • Flexible cradle system has the benefit that different combinations of vessels can be used offering maximum versatility.
Specifications
Product Type Platform
Dimensions (L x W) 510 x 510 mm
For Use With (Application) For SGM-265-010C
For Use With (Equipment) Sutart™ Orbirtal Shakers SSM1, SSL1