Learn More
Cole-Parmer™ Clip Plate for Rotator Drum
Product Code. 12043324
Click to view available options
For Use With (Equipment):
For 10 Tubes
For 12 Tubes
For 7 Tubes
For 8 Tubes
Unit Size:
Pack of 2
Product Code. 12043324
Brand: Cole-Parmer™ 5160092
Only null left
Add to basket
Add to basket
Optimize performance of your DR-200 Series Rotating Shaker System with this Clip Plate for Rotator Drums.
  • Change your rotator's capacity with interchangeable drums
  • Clip plates hold various size tubes
Drums are compatible with Cole-Parmer™ DR-200 Series Rotating Shaker System Drives (sold separately). Use drive unit in conjunction with a wide range of drums and clip plates designed to take different sizes and types of vessels.
Specifications
Product Type Tube Rack
Holds Tubes (16 mm)
Positions 10
For Use With (Equipment) For 10 Tubes
Diameter (Metric) Opening 16 mm