Learn More
Cole-Parmer™ Clip Plate for Rotator Drum
Click to view available options
For Use With (Equipment):
For 10 Tubes
For 12 Tubes
For 7 Tubes
For 8 Tubes
Unit Size:
Pack of 2
Description
- Change your rotator's capacity with interchangeable drums
- Clip plates hold various size tubes
Drums are compatible with Cole-Parmer™ DR-200 Series Rotating Shaker System Drives (sold separately). Use drive unit in conjunction with a wide range of drums and clip plates designed to take different sizes and types of vessels.
Specifications
Specifications
| Product Type | Tube Rack |
| Holds | Tubes (16 mm) |
| Positions | 10 |
| For Use With (Equipment) | For 10 Tubes |
| Diameter (Metric) Opening | 16 mm |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur