Learn More
Cole-Parmer™ Stuart™ SM5/SWIRL Mini Stirrer
Product Code. 12937216
Change view
Click to view available options
Model:
SM5/Swirl Fascia
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
12937216 SM5/Swirl Fascia Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 12937216 Supplier Cole-Parmer™ Supplier No. 0480121
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Regulatory: This product is under eco-contribution tax.
Powerful and lightweight small-footprint magnetic stirrer. Stuart™ SM5/SWIRL Mini Stirrer is made from robust polypropylene and polycarbonate.
  • Lightweight stirrer.
  • Capable of stirring up to 1 L.
  • Universal power adaptor.
  • BioCote antimicrobial protection.
  • Robust polypropylene with a chemically resistant polycarbonate top.
Specifications
Speed 350 to 2000 rpm
Top Plate Material Polypropylene
Voltage 120/230 V
Dimensions (L x W x H) 66 x 143 x 143 mm
Height (Metric) 66 mm
Width (Metric) 143 mm
Electrical Requirements 120-230V, 50-60Hz
Frequency 50/60 Hz
Model SM5/Swirl Fascia
Type Magnetic Mini Stirrer
Weight (Metric) 500 g