Learn More
Cole-Parmer™ Stuart™ Wall Supports
Click to view available options
:
For use with Bibbyjet Pro Pipette Controller PC2000/2001
Description
- Lightweight and ergonomic
- Powerful enough to fill a 50 mL pipette in just 10s
- Battery sufficient to give a full day's work before recharging - LED display shows when battery is low
- Pipette holder fully autoclavable
- Venting feature allows air sucked from the pipette to be discharged from the front of the unit, avoiding corrosion of internal mechanisms
Specifications
Specifications
| Type | Wall Support |
| For Use With (Equipment) | Bibbyjet Pro Pipette Controller PC2000/2001 |