Learn More
Hoefer™ SUB10 Mini-Plus Submarine Combs
Accessories for SUB10 Mini-Plus Submarine
Brand:  Hoefer™ SUB10-C1-8
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12615726
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Ranges from 8, 10, 12, 16 and 20
  • 1, 1.5 and 2 mm Thickness
  • Size ranges from 3 to 9 mm Well Width
  • Min. 12, max. 80 μL Sample Volume
Specifications
Specifications
SUB10 Mini plus Submarine Unit
8
0.1 cm
0.35 in.
0.19 in.
Comb
0.04 in.
40 μL
0.9 cm
0.5 cm