Learn More
Hoefer™ SUB10 Mini-Plus Submarine Combs
Accessories for SUB10 Mini-Plus Submarine
Brand: Hoefer™ SUB10-C2-8
Additional Details : Weight : 0.02000kg
Description
- Ranges from 8, 10, 12, 16 and 20
- 1, 1.5 and 2 mm Thickness
- Size ranges from 3 to 9 mm Well Width
- Min. 12, max. 80 μL Sample Volume
Specifications
SUB10 Mini plus Submarine Unit | |
8 | |
0.2 cm | |
0.35 in. | |
0.19 in. |
Comb | |
0.07 in. | |
80 μL | |
0.9 cm | |
0.5 cm |