Learn More
Suberic acid, 99%
Description
Suberic acid is used in drug synthesis and in the production of plastics. It is also used in the preparation of reduction-sensitive micelles, which affects their cellular uptake. Further, it is used in the delivery of anticancer drugs. In addition, it is used in the fluorescent detection of amidinium-carboxylate and amidinium formation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suberic acid is used in drug synthesis and in the production of plastics. It is also used in the preparation of reduction-sensitive micelles, which affects their cellular uptake. Further, it is used in the delivery of anticancer drugs. In addition, it is used in the fluorescent detection of amidinium-carboxylate and amidinium formation.
Solubility
Soluble in water,dimethyl sulfoxide and methanol. Slightly soluble in ether. Insoluble in chloroform.
Notes
Incompatible with strong oxidizing agents, reducing agents and bases.
Specifications
Specifications
| Melting Point | 141°C to 144°C |
| Boiling Point | 230°C (15 mmHg) |
| Flash Point | 203°C (397°F) |
| Quantity | 100 g |
| Beilstein | 1210161 |
| Merck Index | 14,8862 |
| Solubility Information | Soluble in water,dimethyl sulfoxide and methanol. Slightly soluble in ether. Insoluble in chloroform. |
| Formula Weight | 174.2 |
| Percent Purity | 99% |
| Chemical Name or Material | Suberic acid |
Safety and Handling
- Suberic acid
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Octanedioic acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.