missing translation for 'onlineSavingsMsg'
Learn More

Suberoylanilide Hydroxamic Acid, TRC

Product Code. 30411029
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30411029 100 mg
30411857 250 mg
30424255 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30411029 Supplier Toronto Research Chemicals Supplier No. TRCS688700100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149647-78-9
Molecular Formula C14 H20 N2 O3
Molecular Weight (g/mol) 264.32
Synonym N1-Hydroxy-N8-phenyl-octanediamide, Octanediamide, N-hydroxy-N'-phenyl- (9CI), N-Hydroxy-N'-phenyloctanediamide, N1-Hydroxy-N8-phenyloctanediamide, SAHA, Suberoyl anilide bishydroxamide, Suberoylanilide hydroxamic acid, Vorinostat, Zolinza
IUPAC Name N'-hydroxy-N-phenyloctanediamide
SMILES ONC(=O)CCCCCCC(=O)Nc1ccccc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H20N2O3/c17-13(15-12-8-4-3-5-9-12)10-6-1-2-7-11-14(18)16-19/h3-5,8-9,19H,1-2,6-7,10-11H2,(H,15,17)(H,16,18)
Formula Weight 264.1474
Percent Purity >95%
Chemical Name or Material Suberoylanilide Hydroxamic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.