missing translation for 'onlineSavingsMsg'
Learn More

Succinate, 0.2M buffer soln., pH 5.0, Thermo Scientific™

Catalog Number p-7042813
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy
150-90-3
C4H4Na2O4
162.052
MFCD00002789
ZDQYSKICYIVCPN-UHFFFAOYSA-L
9020
CHEBI:63675
disodium;butanedioate
C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]
This item is not returnable. View return policy
150-90-3
C4H4Na2O4
162.052
MFCD00002789
ZDQYSKICYIVCPN-UHFFFAOYSA-L
9020
CHEBI:63675
disodium;butanedioate
C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Concentration 0.2 M
pH 5.0
Physical Form Liquid
Quantity 250 mL
Odor Odorless
Chemical Name or Material Succinate buffer soln.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'