Learn More
Sucrose Brix/RI Standard, UKAS ISO 17025 / ISO 17034 Certified, Sucrose (11.20 °Brix / 1.34968 RI) Standard, 15 mL, Paragon Scientific™
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Primary level refractive index standards traceable to National Standards
- Manufactured in accordance with ICUMSA Methods
- Supplied in tamper evident packs with 6 weeks shelf life
Specifications
Specifications
| Quantity | 15 mL |
Nominal values quoted. Certified Values will vary from batch to batch.