Learn More
Sucrose Brix Standard Solution, 40%, Reagecon™
Product Code. 12633166 Shop All Reagecon Products
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
Product Code. 12633166
Brand: Reagecon BS40