Learn More
Erlab Suction Protection Grill
To avoid any object suctions and therefore to offer the best work comfort, ERLAB offers this suction protection grill as standard equipment.
Brand:  Erlab NORT000384
Product Code. 17786594
  • 19.05€ / Each
Description
  • Grill placed at the entrance of the extraction duct to avoid suction of rags, wipes or any light material by the fan.