missing translation for 'onlineSavingsMsg'
Learn More

Sulfamic Acid, ≥99%, Reagent Grade, Honeywell™

Catalog Number p-7120846
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
5329-14-6
H3NO3S
97.088
MFCD00011603
IIACRCGMVDHOTQ-UHFFFAOYSA-N
amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
5987
CHEBI:9330
sulfamic acid
NS(=O)(=O)O
This item is not returnable. View return policy
5329-14-6
H3NO3S
97.088
MFCD00011603
IIACRCGMVDHOTQ-UHFFFAOYSA-N
amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
5987
CHEBI:9330
sulfamic acid
NS(=O)(=O)O

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Appearance Crystalline powder
Color Colorless
Melting Point 205°C
Density 2.1g/cm3
Packaging Poly Bottle
Quantity 5 g
Linear Formula NH2SO3H
UN Number UN2967
Formula Weight 97.09g/mol
Percent Purity ≥99%
Grade ReagentPlus™
Chemical Name or Material Sulfamic Acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'