missing translation for 'onlineSavingsMsg'
Learn More

Sulfamic acid, Honeywell Fluka™

Catalog Number 15655310
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
This item is not returnable. View return policy
5329-14-6
H3NO3S
97.088
MFCD00011603
IIACRCGMVDHOTQ-UHFFFAOYSA-N
amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
5987
CHEBI:9330
sulfamic acid
NS(=O)(=O)O
This item is not returnable. View return policy
5329-14-6
H3NO3S
97.088
MFCD00011603
IIACRCGMVDHOTQ-UHFFFAOYSA-N
amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
5987
CHEBI:9330
sulfamic acid
NS(=O)(=O)O
Quantity 2.5 kg
Linear Formula NH2SO3H
UN Number UN2967
Formula Weight 97.09g/mol
Percent Purity 99.3 to 100.3% (dry basis)
Chemical Name or Material Sulfamic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'