missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Sulfate Test Kits

Product Code. 10472151
Click to view available options
Unit Size:
Each
Pack of 100
Pack of 25
Range:
0 to 70 mg/L
100 to 1000 mg/L
5 to 250 mg/L
This item is not returnable. View return policy

Product Code. 10472151

Brand: WTW™ 250449

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Get accurate test results for your ground water, building materials, and soils with the photometric test kits for measurement of Ammonium NH4.

  • Photometric test kit for measurement of Ammonium NH4
  • Variety of instrumentation for measuring CombiCheck Ammonium (NH4)
  • CombiCheck Ammonium (NH4) cell test kit
  • For use with PhotoLab® S6, PhotoLab® S12, PhotoLab® Spectral, Photoflex photometers
  • For ground water, building materials, soils
TRUSTED_SUSTAINABILITY

Specifications

Analytes Iron(III) thiocyanate
For Use With (Application) For ground water, building materials, soils
For Use With (Equipment) For PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral photometers
Includes Certificates
No. of Tests 90
Sufficient For 25 Tests
Test Method Photometric
Type CombiCheck Chloride (Cl) cell test kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.