Learn More
Witeg™ Sulfonation Flasks
An excellent vessel for use in pharmaceutical applications.
133.00€ - 135.00€
Specifications
Material | Glass |
---|---|
For Use With (Application) | Pharmaceutical |
Neck Style | 4 side necks, 1 center |
Product Type | Sulfonation flask |
Socket Joint | CN ST 29/32, SN ST 19/26 |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11817800
|
Witeg™
0 850 032 |
200 mL |
135.00€
Each |
Estimated Shipment: 04-07-2024
Log in to see stock. |
|||||
11807800
|
Witeg™
0 850 031 |
100 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
- Available in either 100 or 200 mL capacities
- With 4 side necks
- Center neck socket ST 29/32
- Side necks socket ST 19/26
Specifications
Glass | |
4 side necks, 1 center | |
CN ST 29/32, SN ST 19/26 |
Pharmaceutical | |
Sulfonation flask |