missing translation for 'onlineSavingsMsg'
Learn More

Sulfur, plasma standard solution, Specpure™ S 1000μg/mL, Thermo Scientific Chemicals

Catalog Number p-4782165
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
(NH4)2SO4 in H2O
MFCD00085316
This item is not returnable. View return policy
(NH4)2SO4 in H2O
MFCD00085316

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 mL
Concentration or Composition (by Analyte or Components) Ammonium sulfate: 0.413%; Water: 99.587%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Chemical Name or Material Sulfur, plasma standard solution

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'