missing translation for 'onlineSavingsMsg'
Learn More

Sulphur solution for ICP/MS 10mg/L H2O matrix, Fisher chemical™

Catalog Number 12927413
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Bottle
This item is not returnable. View return policy
7704-34-9
S
32.06
MFCD00085316
NINIDFKCEFEMDL-UHFFFAOYSA-N
5362487
CHEBI:26833
sulfur
[S]
This item is not returnable. View return policy
7704-34-9
S
32.06
MFCD00085316
NINIDFKCEFEMDL-UHFFFAOYSA-N
5362487
CHEBI:26833
sulfur
[S]

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 10mg/L
Packaging Bottle
Quantity 50 mL
Solvent or Matrix H2O
Chemical Name or Material Sulphur
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'