missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MACF1 Recombinant Protein Antigen

Product Code. 18690317 Shop All Bio Techne Products
Click to view available options
:
0.1 mL
This item is not returnable. View return policy

Product Code. 18690317

Brand: Novus Biologicals™ NBP249542PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1 M Urea, pH 7.4.
Gene ID (Entrez) 23499
Molecular Weight (g/mol) MolecularWeight-theroretical: 28 kDa
Quantity 0.1 mL
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Gene Alias microtubule-actin crosslinking factor 1
Gene Symbol MACF1
Inhibitor Properties This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-49542. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL
Research Category Cell Cycle and Replication
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein MACF1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.