Learn More
Thermo Scientific™ SuperBlock™ (TBS) Blocking Buffer
Block membranes, plates and tissue samples with these optimized solutions containing a protein that provides fast and effective blocking for Western blot, ELISA and IHC.
44.58€ - 229.00€
Products 2
Product Code Brand Product Name Quantity Price Quantity & Availability  
Product Code Brand Product Name Quantity Price Quantity & Availability  
10016723 View Documents Thermo Scientific™
37535
SuperBlock (TBS) Blocking Buffer 1L 229.00€
1 L View alternative products
Save Up to 
»
In stock, ready to ship: 32
Add to basket
13444299 View Documents Thermo Scientific™
37581
SuperBlock (TBS) Blocking Buffer 100mL 44.58€
100mL View alternative products
Save Up to 
»
In stock, ready to ship: 11
Add to basket
Description
Description
Blocking microplates, membranes or tissues with a SuperBlock Buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.
Highlights:
  • Fast – block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
  • Flexible – guaranteed to be biotin-free for use with streptavidin system
  • Convenience – available in multiple buffer formats (in PBS or TBS with or without Tween™-20 detergent) and package sizes (1L, 5L and dry-blend pouches)
  • Low background – the non-serum protein solution yields a high signal-to-noise ratio
  • Stable – store buffer at 4°C for one year; store blocked plates dry for up to 12 months