Learn More
Fisherbrand™ SureOne™ Large Orifice Filtered Pipette Tips
GREENER_CHOICE
Product Code. 11947744 Shop All Fisherbrand Products
Change view
Click to view available options
Unit Size:
Pack of 960
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
11947744 Pack of 960
11977744 Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11947744
Only null left
Add to basket
Edge
Add to basket
Using Fisherbrand™ Sure One™ large orifice pipette tips can help maintain the integrity of genomic DNA (gDNA) and reduce the risk of cell shearing during pipetting. With a 1.5 mm inner diameter tip orifice helps reduce shearing forces and are best suited for handling viscous or particulate solutions.
  • Large orifice, 1.5 mm inner diameter, helps reduce shearing forces
  • Low Retention, hydrophobic feature provides maximum sample recovery
  • Non-sterile products are certified to be free of RNase, DNase, DNA and Pyrogen
  • Sterilized products are certified to be free of RNase, DNase, DNA, Pyrogen/Endotoxin, PCR Inhibitors, ATP and Bioburden
  • Universal Fit, compatible with most popular brands of pipettor (optimal fit with Fisherbrand Elite™ pipettor)
Available bulk, rack non-sterile, rack sterile, and filtered rack sterile
TRUSTED_SUSTAINABILITY
Specifications
Filtered Filtered
Sterility Sterile
Color Clear
Length (English) 1.96 in.
Length (Metric) 4.97 cm
Clean Claims Sterilized products are certified to be free of RNase, DNase, DNA, Pyrogen/Endotoxin, PCR Inhibitors, ATP and Bioburden
Features Low Retention
Format Hinged Rack Sterile
Tip Style Genomic/Wide Orifice
Packaging 96 Tips/Rack, 960/Pk., 4800/Cs.
Product Type Filtered Pipette Tip
For Use With (Equipment) Universal Fit, fits most pipettes
Graduated No
Material Polypropylene
Show More Show Less
As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.