Learn More
Thermo Scientific™ SureQuant™ AKT Isoform Relative Quantitation Module
Product Code. 17103169 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 reactions kit
Unit Size:
10 reactions
Product Code. 17103169
Brand: Thermo Scientific™ A40092
Thermo Scientific™ SureQuant™ AKT Isoform Relative Quantitation Module provides a complete quantitation solution for the analysis of key AKT expression using mass spectrometry (MS).
  • Complete - contains validated AQUA HeavyPeptide mix, diluent, matrix, system suitability standard including optimized acquisition and analysis methods
  • Validated - peptides rigorously tested for successful quantitation of each target peptide
  • Multiplex - able to quantitate AKT1, AKT2, and AKT3 isoforms and phosphorylation of AKT
TRUSTED_SUSTAINABILITY
Specifications
Detection Method Mass Spectrometry
For Use With (Equipment) Thermo Scientific™ Q Exactive Mass Spectrometer, Mass Spectrometer
Shipping Condition Approved for shipment on Wet or Dry Ice
Workflow Step Target Selection
Product Line SureQuant
Quantity 10 reactions kit
For Use With (Application) Mass Spectrometry
Type Quantitation Module
Final Product Type Peptides
Starting Material Protein Samples
Sufficient For 10 Reactions