Learn More
Terumo™ SurGuard2™ Hypodermic Safety Needles
Designed with safety in mind, these hypodermic needles include a simple locking mechanism to reduce needle stick injuries. Terumo™ SurGuard2™ Hypodermic Safety Needles can be easily activated using only one hand.
18.50€
Specifications
Includes Safety lock
Product Type Hypodermic Needle
Quantity 800/Pk.
View More Specs
Products 2
Product Code Brand Needle Gauge Price Quantity & Availability  
Product Code Brand Needle Gauge Price Quantity & Availability  
11700805
View Documents Promotion Details
Terumo™
8SG22613MP
26 G This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12661146
View Documents Promotion Details
Terumo™
8SG22516MP
25 G This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description
Description
  • Simple locking mechanism for safety
  • Safety lock can be activated using only one hand
  • Standard hub equally fits to all luer lock and luer slip syringes
Specifications
Specifications
Safety lock
Hypodermic Needle
800/Pk.