Learn More
Pyrex™ SVL™ Stirrer Heads for SVL™ Stirrer Shafts
Click to view available options
Type:
Anchor
Collapsible Impeller
Helix - 6 Blades
Paddle
Diameter (Metric):
54 mm
70 mm
80 mm
90 mm
Unit Size:
Each
Pack of 5
Description
- Designed for use with SVL™ stirrer shafts
- Head simply screws into the end of the shaft
- Manufactured from a PTFE based polymer with the Anchor and Helix types stiffened by a metal core
Specifications
Specifications
| Diameter (Metric) | 70 mm |
| For Use With (Equipment) | SVL reaction vessels |
| Quantity | 1 Each |
| Type | Paddle |