Learn More
Neogen Swab-Sampler
Product Code. 17228520
Encompass
Click to view available options
For Use With (Application):
1 mL Letheen Broth
4 mL Letheen Broth
4 mL Neutralizing Buffer
5 mL Letheen Broth
Unit Size:
Pack of 100
Product Code. 17228520
Brand: Neogen 7000030299
This item requires a price update prior to being ordered.
Request a Price for this item here.
Convenient and quick collection device for environmental sampling and testing over small surface areas.
  • Letheen Broth vial is ready to use for pathogen testing of environmental samples.
  • Swab reaches into corners and hard-to-reach areas for increased accuracy.
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant.
  • Bottles have ample headspace for sample addition and vortexing.
  • Self-contained swab and vial for increased productivity and efficiency.
Specifications
For Use With (Application) 1 mL Letheen Broth
Quantity 100 /Cs.
Product Type Swab