Learn More
Thermo Scientific™ SWAP-IT™ GC-MS Interface System
Product Code. 10702327 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Agilent GC-MS Instruments
DSQ GC-MS, DSQ II GC-MS, PolarisQ GC-MS, ITQ GC-MS Only
ISQ GC-MS, TSQ 8000 GC-MS Systems Only
ISQ GC-MS, TSQ 8000 GC-MS/MS
Includes:
Interface, Nuts, Ferrules, and Accessories
SWAP-IT Interface only
Unit Size:
Each
Product Code. 10702327
Brand: Thermo Scientific™ 60180794
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Make column changeovers quick and easy. Use this GC-MS interface system to switch capillary columns without the need to vent your mass spectrometer (MS).
Make column changeovers quick and easy. The Thermo Scientific™ SWAP-IT™ GC-MS Interface system is a powerful tool that enables capillary column changes without the need to vent your mass spectrometer (MS). It uses a specially deactivated, robust stainless steel transfer capillary as an interface between the analytical column and the MS detector. The small inner diameter of transfer capillary ensures minimum exposure to air and water when the analytical column is disconnected. A helium purge is not required. Finger-tight connections make it quick and easy without the need for tools.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Type GC-MS Interface Kit
Description SWAP-IT GC-MS Interface Kit
Includes Interface, Nuts, Ferrules, and Accessories
For Use With (Equipment) Agilent GC-MS Instruments