Learn More
Sigma Laborzentrifugen™ Swing Out Rotor
Process microplates, PCR plates/tube strips and many commonly used centrifugal filtration and purification devices with these Sigma™ swing out rotors.
Brand:  Sigma Laborzentrifugen™ 11134
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11810320
  • 2540.00€ / Each
Estimated Shipment
29-06-2021
Add to basket
Description
Description
  • Precision-made from modern, certified high-performance materials
  • Can be operated continuously at maximum speed and +4°C in nearly all Sigma centrifuges
  • Can be used for many cycles with maximum load and at maximum speed, up to 50,000 cycles
Specifications
Specifications
Very high speed
4 x 10 mL Round Bottom Tubes
10,000 rpm
2 sets no. 13134 e.g. 15000, 15039
Swing-out Rotor
10,956 x g
90°