Learn More
Sigma Laborzentrifugen™ Swing Out Rotor
Process microplates, PCR plates/tube strips and many commonly used centrifugal filtration and purification devices with these Sigma™ swing out rotors.
POA
Specifications
Type Swing-out Rotor
View More Specs
Description
Description
  • Precision-made from modern, certified high-performance materials
  • Can be operated continuously at maximum speed and +4°C in nearly all Sigma centrifuges
  • Can be used for many cycles with maximum load and at maximum speed, up to 50,000 cycles
Specifications
Specifications
Swing-out Rotor