Learn More
Simport™ Scientific Swingsette™ Tissue Processing/Embedding Cassette in Quickload™ stacks
Secure specimens during processing with Simport Scientific™ Swingsette™ Tissue Processing/Embedding Cassette, in Quickload™ stacks.
Description
In Quickload™ stacks
- 45° Anterior writing area
- Easy to assemble hinged lids
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.
Certifications
CE-IVD
Specifications
| Acetal | |
| 2000 |