missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation Symmetry C18 Column
Symmetry C18 columns feature monofunctionally bonded C18 ligands on a high purity base-deactivated silica.
Brand: Waters Corporation 186000690
Additional Details : Weight : 0.50000kg
Description
- Symmetry columns have the tightest column specifications in the industry giving resulting highly reproducible chromatographic results.
- Symmetry columns continues to set the standard for HPLC drug assays, giving you confidence in the long-term compliance of your HPLC methods.
- No need to compromise on peak shape, selectivity, column-to-column and batch-to-batch reproducibility, or other critical performance characteristics.
- Available in a variety of particle sizes, chemistries and dimensions.
Specifications
3 mm | |
6000 psi (415 Bar) | |
5 μm | |
1/Ea. | |
Yes |
250 mm | |
Spherical | |
100 Å | |
L1 | |
Column |