Learn More
Syndecan 3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553553
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TPRLVSTATS RPRALPRPAT TQEPDIPERS TLPLGTTAPG PTEVAQTPTP ETFLTTIRDE PEVPVSGGPS GDFELPEEET TQPDTANEVV AVGGAAAKAS SPPGTLPKGA RPGPGLLDNA IDSGSSAAQL PQKSILERK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
Syndecan 3 Polyclonal antibody specifically detects Syndecan 3 in Human samples. It is validated for Immunohistochemistry.Specifications
Syndecan 3 | |
Polyclonal | |
Unconjugated | |
Sdc3 | |
Kiaa0468; mKIAA0468; Neuroglycan; N-syndecan; SDC3; SDCN; syn-3; Synd3; syndecan 1; syndecan 3; syndecan neural type; syndecan proteoglycan 3; syndecan-3; wu:fa97a01 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9672 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75056 | |
Sdc3 | |
Recombinant protein corresponding to Human Syndecan 3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only