Description
- HILED Lighting option for GBOX
- Lighting units: Red, Green, Blue and Infra-Red
- Includes gantry and electronics
HILED, Multiplex. Fluorescence, westerns
Specifications
Specifications
| Product Line | Syngene |
| For Use With (Application) | Fluorescence westerns |
| For Use With (Equipment) | Syngene Gbox |
| Quantity | 1Each |
| Wavelength | Blue, green, red, infrared |
| Product Type | HILEDS |