missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polytetra-fluorethylene Non-sterile Syringe Filter

Fisherbrand™ Polytetra-fluorethylene Non-sterile Syringe Filter is hydrophobic membrane with resistance against most of organic solvents and aggressive liquids.

Brand:  Fisherbrand™

Additional Details : Weight : 0.01000kg

Product Code. 11730126

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »


Description

Description

  • Hydrophobic membrane
  • High thermal and chemical resistance
  • Resistance against most of organic solvents and aggressive liquids
  • Chemical compatibility: pH 1-14
Specifications

Specifications

Bubble point: 0.45μm
Non-sterile
Syringe Filter
PTFE
Hydrophobic
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions