missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ Syringe fixed needle superflex for valco/rheodyne injectors SGE

Fixed needle with LC tip and flexible plunger.

Brand:  Trajan Scientific and Medical™ 002330

Additional Details : Weight : 0.50000kg

Product Code. 12977546

  • 350.00€ / Pack of 6
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Beckman/Altex, Rheodyne, Valco, SSI valves
Specifications

Specifications

SuperfleX (Plunger)
0.17 mm
6/Pk.
LC
22 G
0.028 mm
10 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions