Learn More
Trajan Scientific and Medical™ Syringe Spark Holland LC autosampler 1/4-28 UNF thread 250μL SGE
For HPLC Applications that provide accurate and reproducible injections
Brand:  Trajan Scientific and Medical™ 006683
Additional Details : Weight : 0.50000kg
Product Code. 12917566
  • 134.00€ / Each
Estimated Shipment: 14-06-2024
to see stock.
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Spark Holland instruments
Specifications
Specifications
PTFE (Tip)
250 μL
Spark Holland Promis LC Autosampler
Gas Tight