missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYTOX™ Red Dead Cell Stain, for 633 or 635 nm excitation

Product Code. 10301833
Click to view available options
Quantity:
1 mL, 1000 assays
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 10301833

Brand: Invitrogen™ S34859

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

A simple and quantitative single-step, dead-cell indicator for use with red laser-equipped flow cytometers.

  • A simple and quantitative single-step dead-cell indicator for use with red laser-equipped flow cytometers
  • SYTOX™ Red dead cell stain is a high-affinity nucleic acid stain that easily penetrates cells with compromised plasma membranes but will not cross uncompromised cell membranes
  • After brief incubation with SYTOX™ Red stain, the nucleic acids of dead cells fluoresce bright red when excited with 633 or 635 nm red laser light
  • SYTOX™ Red dead cell stain is distinct from other dead cell probes like 7-AAD and PI, which are excited using 488 nm
  • The emission of SYTOX red stain is limited to one channel with minimal spectral overlap
  • This effectively frees all the channels of the 488 nm laser line
  • Supplied as a 5 μM solution in dimethylsulfoxide (DMSO)
TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Impermeant
Color Red
Concentration 5 μM
Content And Storage Contains 1 vial of SYTOX™ red dead cell stain (5 μM solution in DMSO).

Store in freezer (-5°C to -30°C), and protect from light.
Description SYTOXTM Red Dead Cell Stain, for 633 or 635 nm excitation
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Dead Cell Stain
Dye Type SYTOX™ Red
Emission 658
Excitation Wavelength Range 640 nm
Form Solution
Format Tube(s)
Product Line SYTOX
Quantity 1 mL, 1000 assays
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Sub Cellular Localization Nucleus, Nucleic Acids
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.