Learn More
IKA™ T 50 Digital Dispersing Element with Jet Mixer Head
Shop All IKA Products
Click to view available options
For Use With (Equipment):
S50 N – W 65SK
Unit Size:
Each
Specifications
Specifications
| Product Type | Dispersing Element |
| Volume Range (Metric) | 1 to 50 L |
| Diameter (Metric) | 80 mm |
| Length (Metric) Shaft | 350 mm |
| For Use With (Equipment) | S50 N – W 65SK |
| Model | S 50N–W65-SK |
| Packaging Quantity | 1 Ea. |