missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ T 5601640
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
T 5601640 is a selective LIM kinase 2 (LIMK2) inhibitor; inhibits cofilin phosphorylation in cells that overexpress LIMK2 but not LIMK1. Attenuates the growth of several cancer cell lines. Reduces phospho-cofilin levels and Panc-1 tumor size in a mouse xenograft model.
Specifications
Specifications
| Purity | >99% |
| Quantity | 50 mg |
| Product Type | T 5601640 |
| Molecular Weight (g/mol) | 389.33 |
| Inhibitors | T 5601640 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction