Learn More
T Cell Exhaustion Marker (Flow) Antibody Pack, Novus Biologicals™
Product Code. 18302614 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
9 Vials
Unit Size:
Pack of 9
Product Code. Quantity unitSize
18302614 9 Vials Pack of 9
1 options
Product Code. 18302614
Brand: Novus Biologicals NBP312864
Includes
F0102B: Goat anti-Mouse IgG Secondary Antibody [Phycoerythrin], F0110: Goatanti-Rabbit IgG Secondary Antibody [Phycoerythrin], F0105B: Goat anti-Rat IgGSecondary Antibody [Phycoerythrin], MAB10863-SP, MAB3253-SP, MAB7898-SP, MAB2365R-SP, MAB71263-SP, MAB23191-SP
Antibody Pack
The Human T Cell Exhaustion Marker Sample Pack (Flow) contains antibodies to detect expression of inhibitory receptors associated with T cell exhaustion including PD-1, CTLA-4, TIGIT, TIM-3, LAG-3 and VISTA. Immune cells expressing these inhibitory receptors are found during chronic viral infection and within the tumor microenvironment of many solid tumors. Also included are three PE-Conjugated secondary antibodies for fluorescent detection by flow cytometry.All antibodies have been validated for use in flow cytometry applications.
TRUSTED_SUSTAINABILITY
Specifications
Antigen T Cell Exhaustion Marker (Flow)
Applications Flow Cytometry
Conjugate Unconjugated
Formulation See individual datasheets
Gene Alias exhaustion markers, immune cell exhaustion, immune checkpoint markers, inhibitory receptors, T cell exhaustion
Immunogen See individual datasheets
Quantity 9 Vials
Regulatory Status RUO
Content And Storage Storage is content dependent.