missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TAC
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TAC is a telomerase reverse transcriptase (TERT) activator. TAC upregulates TERT gene transcription via the MEK/ERK/AP-1 cascade. In primary human cells and naturally aged mice, TAC enhances telomere synthesis, reduces tissue aging markers, cellular senescence, and inflammatory cytokines. In the brain, TAC induces neurogenesis, reduces neuroinflammation, and preserves cognitive function.
Specifications
Specifications
| Chemical Name or Material | N-(3-Chloro-4-fluorophenyl)-4-fluoro-3,5-dimethylbenzenesulfonamide |
| CAS | 666699-46-3 |
| Quantity | 50 mg |
| Target | TERT |
| Molecular Formula | C14H12ClF2NO2S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction