missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TACSTD2 (Human) Recombinant Protein

Human TACSTD2 full-length ORF ( AAH09409, 27 a.a. - 323 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00004070-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157501

  • 528.00€ / 25µg
Estimated Shipment: 10-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: HTAAQDNCTCPTNKMTVCSPDGPGGRCQCRALGSGMAVDCSTLTSKCLLLKARMSAPKNARTLVRPSEHALVDNDGLYDPDCDPEGRFKARQCNQTSVCWCVNSVGVRRTDKGDLSLRCDELVRTHHILIDLRHRPTAGAFNHSDLDAELRRLFRERYRLHPKFVAAVHYEQPTIQIELRQNTSQKAAGDVDIGDAAYYFERDIKGESLFQGRGGLDLRVRGEPLQVERTLIYYLDEIPPKFSMKRLTAGLIAVIVVVVVALVAGMAVLVITNRRKSGKYKKVEIKELGELRKEPSL
Specifications

Specifications

AAH09409
tumor-associated calcium signal transducer 2
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TACSTD2
GST
4070
Wheat germ expression system
25 μg
EGP-1, GA733, GA733-1, M1S1, TROP2
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions