Learn More
TAE Buffer, 10X, Serva Electrophoresis™
Product Code. 11752437
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11752437 10 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11752437 Supplier Serva Electrophoresis Supplier No. 4255304
Specifications
Chemical Name or Material TAE Buffer
Color Colorless
Certificate of Analysis 1907/2006/EC
Concentration 10X
For Use With (Application) Electrophoresis of nucleic acids
pH 8.5
Physical Form Solution
Quantity 10 L
Formulation 48.46g/L (0.4M) Tris, 3.72g/L(0.01M) EDTA-Na2-salt, 12.01g/L (0.2M) Acetic acid
Odor Odorless
Solubility Information Soluble