Learn More
TAE Buffer, 50X, Serva Electrophoresis™
Product Code. 11742437
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
Unit Size:
Each
Product Code. Quantity unitSize
11742437 1 L Each
1 options
Product Code. 11742437
Brand: Serva Electrophoresis 4254901
Specifications
Chemical Name or Material TAE Buffer
Color Colorless
Certificate of Analysis 1907/2006/EC
Concentration 50X
For Use With (Application) Electrophoresis of nucleic acids
pH 8.3
Physical Form Solution
Quantity 1 L
Formulation 242.3g/L Tris, 18.6g/L EDTA-Na2-salt, 60.05g/L Acetic acid
Odor Odorless
Solubility Information Soluble
Show More Show Less
compliance-icons
Product Identifier
  • TAE buffer (50x)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.