missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAG-72 Antibody (rB72.3), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08565UV
491.20 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
TAG-72 Monoclonal antibody specifically detects TAG-72 in Human, Rat, Bovine, Canine, Hamster samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
TAG-72 | |
Monoclonal | |
DyLight 350 | |
CA 72-4, Protein TAG-72, tag 72, tag-72 Protein TAG-72 | |
Membrane-enriched fraction of a human breast carcinoma liver metastasis | |
100 μg | |
Tumor Biomarkers | |
Human, Rat, Bovine, Canine, Hamster | |
Purified |
Immunohistochemistry (Paraffin) | |
rB72.3 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction